Learn More
TARGETMOL CHEMICALS INC PF-02413873 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T28362
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-02413873 is a competitive antagonist of nonsteroidal progesterone receptor with a Ki of 2.6 nM. PF-02413873 can be used in studies about the treatment of gynecological conditions such as endometriosis. purity: 99%
Catalog No. 50-257-7039
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.