Learn More
Apexbio Technology LLC PF-04620110 1109276-89-2 5mg
SDP
Supplier:  Apexbio Technology LLC A43845
Encompass_Preferred
PF-04620110 (CAS 1109276-89-2) is a selective and orally bioavailable inhibitor of diacylglycerol acyltransferase 1 (DGAT-1) an enzyme that catalyzes the final step in triglyceride biosynthesis Inhibition of DGAT-1 by PF-04620110 has been demonstrated with IC50 values of 19 nM in human cell assays and 8 nM in HT-29 cells The compound exhibits over 100-fold selectivity for DGAT-1 compared to other lipid-metabolizing enzymes including human DGAT-2 ACAT-1 ACAT-2 and various monoacylglycerol acyltransferases PF-04620110 is utilized in biomedical research to study triglyceride metabolism and the physiological roles of DGAT-1
Catalog No. 50-000-94782
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.