Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 04628935
GREENER_CHOICESDP

Catalog No. 63475
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-475 5 mg
63-472-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-475 Supplier Tocris Bioscience™ Supplier No. 6347/5

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent ghrelin receptor inverse agonist

PF 04628935 is a potent ghrelin receptor inverse agonist (IC50 = 4.6 nM). Orally bioavailable and exhibits reasonable brain penetration.

Specifications

Chemical Name or Material 1-[2-[[2-Chloro-4-(2H-1,2,3-triazol-2-yl)phenyl]methyl]-2,7-diazaspiro[3.5]non-7-yl]-2-(2-methylimidazo[2,1-b]thiazol-6-yl)ethanone
CAS 1383719-97-8
Assay Percent Range >98%
Quantity 5 mg
Molecular Formula C24H26ClN7OS
Formula Weight Observed MW: 496.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.