Learn More
Apexbio Technology LLC PF-04929113 (SNX-5422) 908115-27-5 50mg
SDP
Supplier:  Apexbio Technology LLC A406550
Encompass_Preferred
PF-04929113 (SNX-5422) [CAS 908115-27-5] is an orally bioavailable water-soluble prodrug of PF-04928473 designed as an inhibitor of heat shock protein 90 (Hsp90) Following oral administration PF-04929113 is rapidly absorbed and converted to its active form PF-04928473 This active metabolite facilitates degradation of HER2 client proteins and demonstrates pronounced antitumor effects in xenograft models including BT-474 and H1650 with minimal observed toxicity at effective doses PF-04928473 additionally suppresses signaling pathways such as p-ERK and p-Akt and reduces angiogenesis markers supporting its use in research on cancer biology and anti-angiogenic strategies
Catalog No. 50-000-95441
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.