Learn More
Medchemexpress LLC HY-120874 5mg Medchemexpress, PF-06372865 CAS:1614245-70-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1208745MG
Encompass_Preferred
Medchemexpress, HY-120874 5mg PF-06372865 CAS:1614245-70-3 PF-06372865 is an orally active, α2/α3/α5 subtype-selective GABAA positive allosteric modulator (PAM). PF-06372865 is a high affinity ligand at GABAA receptors containing α1/α2/α3/α5 subunits (Kis of 2.9 nM, 21 nM, 134 nM for α2, α1 PAM, α2 PAM, respectively), with low affinity for α4/α6 subunits. PF-06372865 can across the blood-brain barrier (BBB). PF-06372865 has anxiolytic activity and has the potential for epilepsy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0954
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.