Learn More
Medchemexpress LLC PF-06380101 (Aur0101), an auristatin microtubule inhibitor, is a cytotoxic Dolastatin 10 analogue. PF-06380101 (Aur0101), 10mM in DMSO in 1ML, CAS# 1436391-86-4
SDP
Supplier:  Medchemexpress LLC HY1252210MM
Encompass_Preferred
PF-06380101 (Aur0101), an auristatin microtubule inhibitor, is a cytotoxic Dolastatin 10 analogue. PF-06380101 (Aur0101), 10mM in DMSO in 1ML, CAS# 1436391-86-4
Catalog No. NC1956703
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.