Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 06651600 malonate
GREENER_CHOICESDP

Catalog No. 65065
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-065 5 mg
65-062-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 65-065 Supplier Tocris Bioscience™ Supplier No. 6506/5

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective JAK3 inhibitor

PF 06551600 malonate is a potent and selective JAK3 inhibitor (IC50 = 33.1 μM). Exhibits no significant activity against a panel of other kinases including JAK1, JAK2 and TYK2. Inhibits phosphorylation of STAT5, induced by IL-2, IL-4, IL-7 and IL-15. Suppresses Th1 and Th17 T-cell differentiation and function in vitro. Reduces paw swelling in a rat model of arthritis and reduces disease severity in a rat EAE model.

Specifications

Chemical Name or Material 1-[(2S,5R)-2-Methyl-5-(7H-pyrrolo[2,3-d]pyrimidin-4-ylamino)-1-piperidinyl]-2-propen-1-one malate
CAS 2140301-97-7
Quantity 5 mg
Molecular Formula C15H19N5O.C3H4O4
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.