Learn More
Medchemexpress LLC PF-06726304 acetate | 1616287-82-1 | MFCD31563584 | >98.0% | 506.38 | C22H21Cl2N3O3 · C2H4O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10368225MG
Encompass_Preferred
PF-06726304 acetate is a potent and selective inhibitor of the histone methyltransferase EZH2, reported to inhibit wild-type and Y641N mutant EZH2 and to reduce tumor growth in xenograft models. It is supplied as a powder, with high HPLC purity and solubility in DMSO and ethanol for biochemical and cellular studies.
  • Potent EZH2 inhibition in biochemical assays.
  • Active against Y641N mutant EZH2.
  • Suitable for biochemical, cellular, and in vivo research.
  • High HPLC purity for reproducible results.
  • Powder form soluble in common organic solvents (DMSO, ethanol).
Catalog No. 50-002-44599