Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 5006739
GREENER_CHOICESDP

Catalog No. 63465
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-465 5 mg
63-462-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-465 Supplier Tocris Bioscience™ Supplier No. 6346/5

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent CK1δ/ε inhibitor

PF 5006739 is a potent casein kinase 1δ/ε inhibitor (IC50 values are 3.9 and 17 nM, respectively). Mediates circadian rhythm phase-delaying effects in vivo. Attenuates opioid drug seeking behavior in addiction in vivo models.

Specifications

Chemical Name or Material 4-[4-(4-Fluorophenyl)-1-[1-(3-isoxazolylmethyl)-4-piperidinyl]-1H-imidazol-5-yl]-2-pyrimidinamine
CAS 1293395-67-1
Assay Percent Range >98%
Quantity 5 mg
Molecular Formula C22H22FN7O
Formula Weight Observed MW: 419.46
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.