Learn More
Medchemexpress LLC HY-107778 10mg Medchemexpress, PF-9366 CAS:72882-78-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10777810MG
Encompass_Preferred
Medchemexpress, HY-107778 10mg PF-9366 CAS:72882-78-1 PF-9366 is a human methionine adenosyltransferase 2A (Mat2A) inhibitor, with an IC50 of 420 nM and a Kd of 170 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8780
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.