Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brooks Automation PF350A/PF350D Compressor

Catalog No. PSUA122 Shop All Brooks Automation Products
Change view
Click to view available options
Voltage:
110 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
PSUA122 110 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PSUA122 Supplier Brooks Automation Supplier No. PSUA122
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benchtop robotic workstation designed for post and pre-analytical management of specimen tubes

The PathFinder 350A:
  • Foil-sealing of tubes: the system can apply a laminated foil cap (heat welded) to tubes, enabling safe storage, transport or freezing (down to -80°C) without risk of leakage
  • Barcode tracking and storage: Each storage rack is barcode-coded, and the system records tube location, enabling retrieval
  • Reduces repetitive strain / injury from manual capping and reduces human error in archiving
The PathFinder 350D:
  • Automates decapping of primary tubes from all major tube manufacturers (both screw‐cap and plug‐cap types) and subsequent sorting
  • Reduces manual handling, reduces risk of repetitive strain injuries, reduces biohazard exposure
  • Tube barcode reading, routing logic, and sorting into analyzer racks or storage, aligned to the labs LIS or workflow

Specifications

Type Compressor
For Use With (Equipment) PathFinder 350A/350D
Voltage 110 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.