Learn More
Siemens Healthineers PFA-100 Collagen/ADP Cartridges (US)
Catalog No. 23044642 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
20 Cartridges
Catalog No. Quantity
23-044-642 20 Cartridges
1 options
This item is not returnable. View return policy
Catalog No. 23-044-642 Supplier Siemens Healthineers Supplier No. 10445699
Add to Cart
Add to Cart
This item is not returnable. View return policy
To aid in the detection of platelet dysfunction in citrated human whole blood.
The Dade™ PFA Collagen/ADP (Col/ADP) test cartridge is used to indicate if an abnormal result obtained with the Col/EPI test cartridge may have been caused by the effect of ASA or medications containing ASA.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Kit
For Use With (Application) Platelet Testing
Quantity 20 Cartridges