Learn More
Siemens Healthineers PFA-100 Collagen/EPI Cartridges (US)
Catalog No. 23044641 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
20 Cartridges
Catalog No. Quantity
23-044-641 20 Cartridges
1 options
This item is not returnable. View return policy
Catalog No. 23-044-641 Supplier Siemens Healthineers Supplier No. 10445697
Add to Cart
Add to Cart
This item is not returnable. View return policy
To aid in the detection of platelet dysfunction in citrated human whole blood.
The Dade™ PFA Collagen/EPI (Col/EPI) test cartridge is the primary cartridge used to detect platelet dysfunction induced by intrinsic platelet defects, VWD, or exposure to platelet-inhibiting agents.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Kit
For Use With (Application) Platelet Testing
Quantity 20 Cartridges