Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ VITLAB™ PFA Narrow-Mouth Reagent Bottles

Catalog No. 14379060 Shop All BrandTech Scientific Products
Change view
Click to view available options
Capacity (Metric):
50 mL
250 mL
500 mL
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14-379-060 500 mL
14-379-061 50 mL
14-379-059 250 mL
14-379-057 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-379-060 Supplier BRANDTECH™ VITLAB™ Supplier No. 108397
Add to Cart
Add to Cart

VITLAB PFA reagent bottles are suitable for trace analysis work and long term storage of standards.

  • Leach-resistant bottles for long-rerm storage of standards
  • Screw caps have a molded sealing ring for a positive seal
TRUSTED_SUSTAINABILITY

Specifications

Capacity 500mL
Capacity (Metric) 500 mL
Capacity (English) 16.9 oz.
Color Translucent
Diameter (English) 2.99 in.
Material PFA
Sterility Non-sterile
Neck Finish S28
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Narrow, Threaded
Shape Sloped Shoulders
Height (Metric) 19 cm
Product Type Reagent Bottle
Height (English) 7.48 in.
Closure Material PFA
Diameter (Metric) 7.6 cm
Closure Type Screw Cap
Autoclavable Autoclavable
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.