Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Verispec™ PFAS Standards, Certified Reference Material
SDP

Catalog No. 01341377 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Chemical Name or Material:
N-Ethyl-N-(2-hydroxyethyl)-PFOS
PFOS tetraethylammonium salt
Perfluorooctane sulfonic acid
Perfluorooctanesulphonamide
Potassium nonafluoro-1-butanesulfonate
Molecular Weight (g/mol):
338.18 g/mol
499.14 g/mol
500.12 g/mol
571.25 g/mol
629.37 g/mol
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material Molecular Weight (g/mol)
01-341-377 N-Ethyl-N-(2-hydroxyethyl)-PFOS 571.25 g/mol
01-341-375 PFOS tetraethylammonium salt 629.37 g/mol
01-341-378 Perfluorooctanesulphonamide 499.14 g/mol
01-341-379 Perfluorooctane sulfonic acid 500.12 g/mol
01-341-384 Potassium nonafluoro-1-butanesulfonate 338.18 g/mol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-341-377 Supplier Verispec™ Supplier No. RV020008100CM
Add to Cart
Add to Cart

Manufactured and tested in an ISO 17025 and 17034 accredited facility for applications in environmental testing, food safety, water quality, and more.

  • Available in convenient 100 mg quantities for customizable calibration standards
  • Supports environmental testing, drinking and wastewater analysis, food safety, and industrial quality control.
  • Ensure compliance with key methods:

EPA: 533, 537.1, 1633, 8327

ASTM: D7968, D7979, D8421, D8535

  • High-quality sample preparation solutions, solvents, and mobile phase reagents
  • Minimize contamination risks to enhance accuracy in sensitive applications
  • Contact Us for customs or ready to use standards

Specifications

Chemical Name or Material N-Ethyl-N-(2-hydroxyethyl)-PFOS
CAS 1691-99-2
Quantity 100 mg
Molecular Weight (g/mol) 571.25 g/mol
Percent Purity Lot Specific
Physical Form Solid or Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.