Learn More
Promega pFC32K Nluc CMV-neo Flexi™ Vector
Catalog No. PRN1341 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
PRN1341 20 μg
1 options
Catalog No. PRN1341 Supplier Promega Supplier No. N1341
Generate C-Terminal Fusions to NanoLuc™ Luciferase Reporter. The pFC32A and pFC32K Nluc CMV-neo Flexi™ Vectors use a directional cloning method based on two rare-cutting restriction enzymes, SgfI and PmeI, and contain a mammalian selectable marker to create a stable line.
  • Generate N- or C-Terminal Fusions to NanoLuc(R) Luciferase Reporter
  • Insert protein of interest using the Flexi(R) Vector System
  • Select options for N- or C-terminal NanoLuc(R) fusions and antibiotic resistance (ampicillin or kanamycin)
  • Bright NanoLuc(R) reporter enables endogenous expression levels of fusion proteins for biologically relevant results
Specifications
Quantity 20 μg
Content And Storage -30°C to -10°C