Learn More
Medchemexpress LLC (E)-1-pyridin-4-yl-3-[7-(trifluoromethyl)quinolin-2-yl]prop-2-en-1-one | 1462249-75-7 | 98.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY122031MG
Encompass_Preferred
PFK-158 is a small-molecule PFKFB3 inhibitor used in research to study cancer cell metabolism, autophagy, and apoptosis. It reduces glucose uptake, ATP production, and lactate release in cancer cells and has demonstrated antitumor activity in preclinical studies.
  • Potent PFKFB3 inhibitor (IC50 ≈ 137 nM).
  • Reduces glucose uptake, ATP production, and lactate release in cancer cells.
  • Induces apoptosis and autophagy in tumor cell models.
  • Supplied as a dry powder for laboratory research.
  • Typical purity ~98.9% and molecular weight 328.29 g/mol.
Catalog No. 50-002-30343
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.