Learn More
Invitrogen™ PFKFB4 Monoclonal Antibody (OTI1C8)

Description
PFKFB4 Monoclonal Antibody for ICC/IF
Specifications
Specifications
| Antigen | PFKFB4 |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1C8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 1% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | PFKFB4 |
| Gene Accession No. | Q16877 |
| Gene Alias | 6-biphosphatase-4; 6PF-2-K/Fru-2,6-P2ase 4; 6PF-2-K/Fru-2,6-P2ASE testis-type isozyme; 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase-4; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 4; bifunctional enzyme with kinase and biphosphatase activities; C230090D14; Fructose-2,6-bisphosphatase; PFK/FBPase 4; Pfkfb4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.