Learn More
MyBioSource PFKP, Rat (Rattus norvegicus) Recombinant Protein, Phosphofructokinase
SDP
Supplier:  MyBioSource MBS2030312
Encompass_Preferred
MyBioSource offers recombinant rat phosphofructokinase (PFKP) protein for scientific research use. This protein is expressed in E. coli and has a molecular weight of 85 kDa. According to the product details, this reagent can be used in activity assays, antibody production, as an immunogen, and in other life science research applications requiring high purity PFKP. MyBioSource provides COA and SDS-PAGE data to verify the identity and purity of the product. This rat PFKP is supplied lyophilized with no additives.
Catalog No. NC2402117
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.