Learn More
Medchemexpress LLC PFM01 100mg | 1558598-41-6 | 293.4 g/mol | C14H15NO2S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY116770100MG
Encompass_Preferred
PFM01 is a small-molecule inhibitor of the MRE11 endonuclease (an N-alkylated Mirin derivative) used in research to modulate DNA double-strand break repair pathways such as non-homologous end joining (NHEJ). It is supplied as a powdered research reagent and commonly distributed in 100 mg amounts.
  • Inhibits MRE11 endonuclease activity
  • Cell-permeable small molecule suitable for cellular studies
  • Used to probe double-strand break repair and NHEJ pathways
  • Supplied as a powder for dissolution in common solvents
  • Typical distributor-reported purity ≥98% (HPLC)
Catalog No. 50-002-27294