Learn More
Promega pFN10A (ACT) Flexi™ Vector
Catalog No. PRC9331 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
PRC9331 20 μg
1 options
Catalog No. PRC9331 Supplier Promega Supplier No. C9331
The CheckMate/Flexi Vector Mammalian Two-Hybrid System provides a means to study suspected interactions between two proteins or domains and can also be used to generate stable cell lines for cell-based assays.
  • Study Mammalian Protein Interactions, Expression and Post-Translational Modifications
  • Confirm, validate and study suspected protein or domain interactions
  • Generate stable cell lines for cell-based assays
  • Enables mammalian protein expression and post-translational modification studies in situ
Specifications
Quantity 20 μg
Content And Storage -30°C to -10°C