Learn More
Promega pFN38K HiBiT CMV-neo Flexi Vector, 20microg
Catalog No. PRN2401 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
PRN2401 20 μg
1 options
This item is not returnable. View return policy
Catalog No. PRN2401 Supplier Promega Supplier No. N2401
This item is not returnable. View return policy
The pFC37K HiBiT, pFN38K HiBiT and pFN39K secHiBiT CMV-neo Flexi Vectors are Flexi Cloning vectors that append the HiBiT tag to the N or C terminus of ORFs; the N terminus for transmembrane or secreted proteins of interest.
  • Vectors for Tagging Proteins with HiBiT for Luminescent Expression Analysis
  • Add an N− or C−terminal HiBiT tag to your protein of interest
  • Generate an N−terminal HiBiT−tagged transmembrane or secreted protein
  • Use with the Flexi(R) Cloning System to transfer ORF insert
Specifications
Content And Storage -30°C to -10°C
Quantity 20 μg