Learn More
Synthware™ PFTE Sleeve
GREENER_CHOICESDP
Catalog No. 31501913 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
29/42
34/45
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-501-913 14/20
31-501-914 19/22
31-501-915 24/40
31-501-916 29/42
31-501-917 34/45
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 31-501-913 Supplier Synthware™ Supplier No. T131420
Add to Cart
Add to Cart
Ribbed with gripping ring
Specifications
Type Ribbed PTFE Sleeve
Material PTFE
For Use With (Application) Organic Synthesis
Standard Taper Joint No. 14/20
Quantity 1 Ea.