Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ PFTE Stopper with Extraction Nut
GREENER_CHOICESDP

Catalog No. 31501907 Shop All Kemtech Products
Change view
Click to view available options
Size:
10/18
14/20
19/22
24/40
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
31-501-907 19/22
31-501-905 10/18
31-501-906 14/20
31-501-908 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 31-501-907 Supplier Synthware™ Supplier No. S531922
Add to Cart
Add to Cart

Stopper has a nylon extraction nut permitting easy removal of the stopper from the glass outer joint.

Specifications

Material PTFE
Color White
Quantity 1 Ea.
Size 19/22
For Use With (Application) Organic Synthesis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.