Learn More
Agilent Technologies PFU ULTRA II FUSION
Supplier: Agilent Technologies 600674
Agilent's PfuUltra II fusion HS DNA polymerase achieves extreme accuracy, high specificity, and long target-length capability while dramatically reducing overall PCR extension times.