Learn More
Miele, Inc. PG 8583 OIL 3AC 208V undercounter labwasher
Supplier:  Miele, Inc. 10254330
Encompass
Designed for simplicity and sustainability, the PG 8583 LDundercounter labwasher reduces energy consumptionand water use in the lab, while making sure mission criticalglassware is analytically clean and ready for the next set of tests. Certified with ACT Label
• High capacity throughput
(up to 128 injector nozzles)
• 128 vials / 98 pipettes
• SimpleLoad modules
• Rear basket docking
• Variable Speed pump
• Auto liquid dispensing
• Multi-stage filter system
• Spray arm monitoring
• ECO Dry - AutoOpen function
• IO/Q Validation
Catalog No. 50-171-2996
Only null left
Add to Cart
Request a Quote