Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGAM1/PGAM2/PGAM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517935 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517935 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517935 Supplier Invitrogen™ Supplier No. PA517935
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

Phosphoglyceric acid mutase is widely distributed in mammalian tissues where it catalyzes the reversible reaction of 3-phosphoglycerate to 2-phosphoglycerate in the glycolytic pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGAM1/PGAM2/PGAM4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PGAM1
Gene Accession No. O70250, P15259, P16290, P18669, P25113, Q8N0Y7, Q9DBJ1
Gene Alias 2310050F24Rik; BPG-dependent PGAM 1; BPG-dependent PGAM 2; CDABP0006; D14Mgh1; dJ1000K24.1; epididymis secretory protein Li 35; GSD10; HEL-S-35; MGC88743; muscle-specific phosphoglycerate mutase; OTTHUMP00000059414; PGAM1; Pgam-1; PGAM2; PGAM3; PGAM4; PGAMA; PGAM-B; PGAMM; PGAM-M; Pgmut; Phosphoglycerate mutase 1; phosphoglycerate mutase 1 (brain); phosphoglycerate mutase 2; phosphoglycerate mutase 2 (muscle); phosphoglycerate mutase 4; phosphoglycerate mutase A, nonmuscle form; phosphoglycerate mutase family 3; phosphoglycerate mutase family member 4; phosphoglycerate mutase isozyme B; Phosphoglycerate mutase isozyme M; phosphoglycerate mutase muscle-specific subunit; phosphoglycerate mutase processed protein; probable phosphoglycerate mutase 4; RP11-452K12.8
Gene Symbols PGAM1, PGAM2, PGAM4
Host Species Goat
Immunogen Synthetic peptide sequence (KAMEAVAAQGKAKK) corresponding to the C-terminus amino acids of PGAM1PGAM2PGAM4 (aa 241-254).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100154776, 100188980, 18648, 24642, 24959, 441531, 5223, 5224, 56012
Target Species Human, Mouse, Rat, Pig
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.