Learn More
PGAM2 Mouse anti-Human, Clone: OTI1C1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503402

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Phosphoglycerate mutase (PGAM) catalyzes the reversible reaction of 3-phosphoglycerate (3-PGA) to 2-phosphoglycerate (2-PGA) in the glycolytic pathway. The PGAM is a dimeric enzyme containing, in different tissues, different proportions of a slow-migrating muscle (MM) isozyme, a fast-migrating brain (BB) isozyme, and a hybrid form (MB). PGAM2 encodes muscle-specific PGAM subunit. Mutations in this gene cause muscle phosphoglycerate mutase eficiency, also known as glycogen storage disease X.Specifications
PGAM2 | |
Monoclonal | |
Unconjugated | |
PGAM2 | |
GSD10, PGAM-M, PGAMM | |
Mouse | |
Affinity Chromatography | |
RUO | |
5224 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1C1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P15259 | |
PGAM2 | |
Full length human recombit protein of human PGAM2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.