Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGBD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541302
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541302 Supplier Invitrogen™ Supplier No. PA541302
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NLPGSLLHTA AYLIQDGSDA ESDSDDPSYA PKDDSPDEVP STFTVQQPPP Sequence homology: Human: 100%.

The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. Anti-PGBD3 belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. Anti-PGBD3 overlaps with the ERCC6 gene on chromosome 10, and pseudogenes of this locus have been found on chromosomes 4, 5 and 12. The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. This gene overlaps with the ERCC6 gene on chromosome 10, and pseudogenes of this locus have been found on chromosomes 4, 5 and 12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGBD3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PGBD3
Gene Accession No. Q8N328
Gene Alias 4732403I04; ATP-dependent helicase ERCC6; C130058G22Rik; Cockayne syndrome protein CSB; CS group B correcting; Csb; DNA excision repair protein ERCC-6; Ercc6; excision repair cross-complementation group 6; excision repair cross-complementing rodent repair deficiency, complementation group 6; FLJ90201; PGBD3; piggyBac transposable element derived 3; piggyBac transposable element-derived protein 3
Gene Symbols PGBD3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PGBD3 (aa 52-101).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 267004
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.