Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGC-1 alpha, Rabbit anti-Mouse, Human, Polyclonal, Millipore Sigma™

Catalog No. ABE86825UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE86825UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE86825UG Supplier MilliporeSigma Supplier No. ABE86825UG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-PGC-1 alpha, Cat. No. ABE868, is a highly specific rabbit polyclonal antibody that targets Peroxisome proliferator-activated receptor gamma coactivator 1-alpha and has been tested for use in Immunocytochemistry and Western Blotting.

Peroxisome proliferator-activated receptor gamma coactivator 1-alpha (UniProt: O70343; also known as PGC-1-alpha, PPAR-gamma coactivator 1-alpha, PPARGC-1-alpha) is encoded by the Ppargc1a (also known as Pgc1, Pgc1a, Ppargc1) gene (Gene ID: 19017) in murine species. PGC-1 alpha is a member of a family of transcription coactivators that plays a central role in the regulation of cellular energy metabolism. It is a homooligomeric transcriptional coactivator for steroid receptors and nuclear receptors that greatly increases the transcriptional activity of PPARG and thyroid hormone receptor on the uncoupling protein promoter. It is expressed in muscles, liver, kidney, brown adipose tissue, heart, and brain. It can regulate key mitochondrial genes that contribute to the program of adaptive thermogenesis and is also reported to play an essential role in metabolic reprogramming in response to dietary availability through coordination of the expression of a wide array of genes involved in glucose and fatty acid metabolism. PGC-1 alpha is known to induce the expression of PPARGC1- and ESRR-induced regulator, muscle 1 (PERM1) in the skeletal muscle in an estrogen-related receptors, -alpha (ESRRA)-dependent manner. PGC-1 alpha is significantly induced in brown adipose tissue and skeletal muscle by exposure to cold and is also shown to be up-regulated in brown adipose tissue of obese leptin-deficient (ob/ob) and leptin-unresponsive (db/db) mice. PGC-1 alpha is also required for oscillatory expression of clock genes, such as ARNTL/BMAL1 and NR1D1, through the co-activation of RORA and RORC, and metabolic genes, such as PDK4 and PEPCK.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGC-1 alpha
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected PGC-1 alpha in A431, HUVEC, and NIH/3T3 cells.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Peroxisome proliferator-activated receptor gamma coactivator 1-alpha;PPAR-gamma coactivator 1-alpha;PPARGC-1-alpha
Gene Symbols Ppargc1a;Pgc1;Pgc1a;Ppargc
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminus region of murine PPAR-gamma coactivator 1-alpha.
Molecular Weight of Antigen ∽100 kDa observed; 90.59 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects PGC-1 alpha in human and murine adipose tissue. it targets an epitope within 21 amino acids from the C-terminus region.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.