Learn More
Promega pGEM™ Express Positive Control Template
Catalog No. PRP2561 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRP2561 10 μg
1 options
Catalog No. PRP2561 Supplier Promega Supplier No. P2561
The pGEM Express Positive Control Template is used to monitor in vitro transcription reactions when using the Riboprobe Systems.
  • Used with Riboprobe(R) Systems to Monitor in vitro Transcription Reactions
  • Can be used with SP6, T7 or T3 RNA polymerase
  • SP6 RNA polymerase produces transcripts of 1,787 and 2,566 bases; T7 RNA polymerase produces transcripts of 1,065 and 2,346 bases; T3 RNA Polymerase produces transcripts of 250 and 1,525 bases
Specifications
Content And Storage -30°C to -10°C
Quantity 10 μg