Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528669
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528669 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528669 Supplier Invitrogen™ Supplier No. PA528669
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, HeLaS3. Predicted reactivity: Mouse (84%), Rat (85%), Pig (87%), Chimpanzee (99%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PGK2 is a form of phosphoglycerate kinase unique to spermatozoa. The autosomal PGK2 gene lacks introns and contains characteristics of a processed gene. The conservation of function in this processed PGK2 gene and its tissue-specific expression in spermatogenesis suggests that it exists as a compensatory response to the inactivation of the X-linked PGK1 gene in spermatogenic cells before meiosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGK2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene PGK2
Gene Accession No. P07205
Gene Alias dJ417L20.2; epididymis secretory protein Li 272; HEL-S-272; Pgk2; Pgk-2; PGKB; PGKPS; phosphoglycerate kinase 2; phosphoglycerate kinase autosomal pseudogene; phosphoglycerate kinase, testis specific; Tcp-2; testicular tissue protein Li 139
Gene Symbols PGK2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PGK2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5232
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.