Learn More
Promega pGL4.54[luc2/TK] Vector
Catalog No. PRE5061 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
PRE5061 20 μg
1 options
Catalog No. PRE5061 Supplier Promega Supplier No. E5061
Vectors are used as controls in dual-luciferase assays where Renilla or NanoLuc luciferase is the experimental reporter.
  • Vectors That Are Co-transfected into Cells as Controls in Dual-Reporter Assays
  • Co-transfect into cells where Renilla or NanoLuc(R) luciferase is the experimental reporter gene
  • SV40, PGK and TK promoter options offer different expression levels suitable for most dual-reporter assays
Specifications
Quantity 20 μg
Content And Storage -30°C to -10°C