Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555132
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555132 Supplier Invitrogen™ Supplier No. PA555132
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SILATRKQSV EDILKDHWQK HGRNFFTRYD YEEVEAEGAN KMMKDLEALM FDRSFVGKQF SANDKVYTVE KADNFEYSD Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Phosphoglucomutases (PGM; EC 5.4.2.2) catalyze the transfer of phosphate between the 1 and 6 positions of glucose. Isozymes of PGM are monomeric, with molecular masses of about 60 kD, and are encoded by several genes, including PGM1. In most cell types, PGM1 isozymes predominate, representing about 90% of total PGM activity. One exception is red cells, where PGM2 (MIM 172000) is a major isozyme (Putt et al., 1993 [PubMed 8257433]). [supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGM1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PGM1
Gene Accession No. P36871
Gene Alias 3230402E02Rik; Aciculin; CDG1T; Glucose phosphomutase 1; glucose phosphomutase 2; GSD14; PGM; PGM 1; PGM 2; Pgm1; Pgm-1; Pgm2; PGM5; PGMRP; PGM-RP; Phosphodeoxyribomutase; phosphoglucomutase 1; phosphoglucomutase 2; phosphoglucomutase 5; phosphoglucomutase isoform 2; phosphoglucomutase isoform1; phosphoglucomutase-1; Phosphoglucomutase-2; phosphoglucomutase-like protein 5; Phosphoglucomutase-related protein; phosphopentomutase; zgc:63718
Gene Symbols PGM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PGM1. Recombinant protein control fragment (Product #RP-94028).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5236
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.