Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGM2 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501726804 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
Catalog No. Quantity
50-172-6804 150 μL
1 options

Catalog No. 50-172-6804

Supplier: Proteintech Group Inc 110221AP150UL

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EWSR1 is a multifunctional protein that is involved in various cellular processes, including gene expression, cell signaling, and RNA processing and transport. The protein includes an N-terminal transcriptional activation domain and a C-terminal RNA-binding domain.

Specifications

Antigen PGM2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PGM2
Gene Accession No. Q96G03
Gene Alias Glucose phosphomutase 2, MSTP006, PGM 2, PGM2, Phosphodeoxyribomutase, phosphoglucomutase 2, Phosphopentomutase
Gene Symbols PGM2
Host Species Rabbit
Immunogen PGM2 Fusion Protein Ag1488
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55276
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.