Learn More
System Biosciences LLC pGreenFire 2.0 NFAT Reporter Virus | >2 x10^6 IFUs
SDP
Supplier:  System Biosciences LLC TR451VAP
Encompass
With the pGreenFire 2.0 NFAT Reporter Lentivector & Virus (pGF2-NFAT-rFluc-T2A-GFP-mPGK-Puro) the core reporter functionality is similar to the original pGreenFire lentivector—NFAT transcriptional response elements (TREs) are placed upstream of a minimal CMV promoter (mCMV) which together drive co-expression of rFLuc and GFP in response to NFAT activity. The result is the ability to quantitatively measure NFAT activity using both fluorescence and luciferase activity.
Catalog No. NC1829697
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.