Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGRP-L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560421
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560421 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560421 Supplier Invitrogen™ Supplier No. PA560421
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VWGTLVLLQR LEPVHLQLQC MSQEQLAQVA ANATKEFTEA FLGCPAIHPR CRWGAAPYRG RPKLLQLPLG FLYVHHTYVP Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 81%.

Peptidoglycan recognition proteins, such as PGRPL, are part of the innate immune system and recognize peptidoglycan, a ubiquitous component of bacterial cell walls.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGRP-L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PGLYRP2
Gene Accession No. Q96PD5
Gene Alias C730002N09Rik; HMFT0141; N-acetylmuramoyl-L-alanine amidase; Peptidoglycan recognition protein 2; peptidoglycan recognition protein L; peptidoglycan recognition protein long; PGLYRP2; Pglyrpl; PGRPL; PGRP-L; TagL; tagL-alpha; tagl-beta; TAGL-like; UNQ3103/PRO10102
Gene Symbols PGLYRP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PGRP-L. Recombinant protein control fragment (Product #RP-99133).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114770
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.