Learn More
Warner Instruments™ PH-6D Platform for Series 20 Chambers, Heated
Catalog No. BTX640290 Shop All BTX Harvard Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Series 20 Microscope Stage Adapters
Catalog No. For Use With (Equipment)
BTX640290 Series 20 Microscope Stage Adapters
1 options
Catalog No. BTX640290 Supplier Warner Instruments™ Supplier No. 640290
The Series 20 Platforms function as a base for the Series 20 Imaging and Recording Chambers and provides the clamping needed to make a seal between a chamber and its bottom glass coverslip.
  • Black anodized aluminum provides good heat conductivity and low stray light reflectance
  • A small hole drilled along platform edge for placement of T1 feedback control thermistor
  • Each Series 20 Platform is compatible with all Series 20 Microscope Stage Adapters
  • Platforms without heating elements can be upgraded later
  • Platforms are machined from aluminum and finished with a black anodize
  • Anodized aluminum platforms provide clamping between our Series 20 chambers and their bottom glass coverslips to create a water tight seal.
Specifications
For Use With (Equipment) Series 20 Microscope Stage Adapters