Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
- Home
- Products
- Buffers and Standards
- Electrochemistry Buffers
- VeriSpec™ pH Buffer, Certified Reference Material
VeriSpec™ pH Buffer, Certified Reference Material
Catalog No. RV1540001A
Shop All Ricca Chemical Company Products

Change view
Click to view available options
Quantity:
500 mL
1 L
Color:
Blue
Colorless
Red
Yellow
pH:
1.00 ±0.01
1.68 ±0.01
10.00 ±0.01
11.00 ±0.01
12.00 ±0.01
12.45 ±0.01
13.00 ±0.01
2.00 ±0.01
3.00 ±0.01
4.00 ±0.01
5.00 ±0.01
6.00 ±0.01
6.86 ±0.01
7.00 ±0.01
8.00 ±0.01
9.00 ±0.01
31 product options available for selection
| Catalog No. | Quantity | Color | pH |
|---|---|---|---|
| RV1540001A | 1 L | Colorless | 6.86 ±0.01 |
| 01-185-028 | 500 mL | Colorless | 1.00 ±0.01 |
| 01-185-027 | 500 mL | Colorless | 1.68 ±0.01 |
| 01-100-270 | 500 mL | Colorless | 2.00 ±0.01 |
| 01-185-026 | 500 mL | Colorless | 3.00 ±0.01 |
| 01-185-025 | 500 mL | Colorless | 4.00 ±0.01 |
| 01-100-272 | 500 mL | Red | 4.00 ±0.01 |
| 01-185-024 | 500 mL | Colorless | 5.00 ±0.01 |
| 01-185-023 | 500 mL | Colorless | 6.00 ±0.01 |
| 01-185-022 | 500 mL | Colorless | 6.86 ±0.01 |
| 01-185-021 | 500 mL | Colorless | 7.00 ±0.01 |
| 01-100-274 | 500 mL | Yellow | 7.00 ±0.01 |
| 01-185-020 | 500 mL | Colorless | 8.00 ±0.01 |
| 01-185-019 | 500 mL | Colorless | 9.00 ±0.01 |
| 01-185-018 | 500 mL | Colorless | 10.00 ±0.01 |
| 01-100-276 | 500 mL | Blue | 10.00 ±0.01 |
| 01-185-017 | 500 mL | Colorless | 11.00 ±0.01 |
| 01-185-016 | 500 mL | Colorless | 12.00 ±0.01 |
| 01-100-277 | 500 mL | Colorless | 12.45 ±0.01 |
| 01-185-015 | 500 mL | Colorless | 13.00 ±0.01 |
| RV1495001A | 1 L | Colorless | 3.00 ±0.01 |
| RV1500001A | 1 L | Colorless | 4.00 ±0.01 |
| 01-100-271 | 1 L | Red | 4.00 ±0.01 |
| RV1505001A | 1 L | Colorless | 5.00 ±0.01 |
| RV1510001A | 1 L | Colorless | 6.00 ±0.01 |
| RV1550001A | 1 L | Colorless | 7.00 ±0.01 |
| 01-100-273 | 1 L | Yellow | 7.00 ±0.01 |
| RV1580001A | 1 L | Colorless | 8.00 ±0.01 |
| RV1590001A | 1 L | Colorless | 9.00 ±0.01 |
| RV1600001A | 1 L | Colorless | 10.00 ±0.01 |
| 01-100-275 | 1 L | Blue | 10.00 ±0.01 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
Description
- No formaldehyde or mercury compounds as preservatives
- No chromium compounds as colorants
- Manufactured and tested in ISO 17025/17034 accredited facilities
- pH 4, 7, and 10 are color coded for easy identification
- Certified traceable to NIST
Specifications
Specifications
| CAS | 7732-18-5, 7558-79-4, 7778-77-0 |
| Color | Colorless |
| Certificate of Analysis | Yes |
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 6.86 ±0.01 |
| Physical Form | Liquid |
| Product Line | VeriSpec |
| Quantity | 1 L |
| Solution Type | pH Buffer |
| Packaging | 1 L Natural Poly |
| Certifications/Compliance | ISO 17025/17034 |
| Grade | Certified |
| Show More Show Less |
ISO 17025/ISO 17034
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?