Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ pH Buffer Solutions (pH 1 to pH 12), 500mL
SCI_ED

Catalog No. S25208C Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
pH 1
pH 10
pH 11
pH 12
pH 2
pH 3
pH 4
pH 5
pH 6
pH 7
pH 8
pH 9
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25208C 500 mL pH 4
S25208K 500 mL pH 12
S25208A 500 mL pH 2
S25208 500 mL pH 1
S25208I 500 mL pH 10
S25208J 500 mL pH 11
S25208B 500 mL pH 3
S25208D 500 mL pH 5
S25208E 500 mL pH 6
S25208F 500 mL pH 7
S25208G 500 mL pH 8
S25208H 500 mL pH 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. S25208C

This product is for educational use - no C of A is available.

Specifications

Chemical Name or Material pH Buffer Solutions (pH 1 to pH 12)
Color Colorless
Density ∽1g/mL
Quantity 500 mL
Packaging pH 4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.