Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies pH, Indicators and Dyes STEM Kit
SDPSCI_ED

Catalog No. S07359 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
40 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S07359 40 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07359 Supplier United Scientific Supplies Supplier No. AISPHKIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Materials for three detailed activities (with model and inquiry investigational paths — a total of four investigational activities), which can accommodate up to 40 students
  • DVD with PDF teacher and student guides
  • Structured, guided and open investigations at introductory, intermediate and advanced levels

Requires

Additional household materials for independent investigations

Introduces students to the chemistry behind plant pigments (the blueberry biopigment cyanidan) and their use as pH indicators and fabric dyes.

  • Students use their knowledge to create their own pH test strips and evaluate them against a commercial type
  • Covers key concepts, including: data analysis, accuracy vs. precision, biopigment structure, pH, pH scale, pH indicators, stains and dyes, acids, bases, acid-base interactions, solutions and self ionization of water
  • Accents the scientific method and independent inquiry featuring hands-on learning materials and exciting interactive digital content that can be accessed on a computer, tablet or mobile device
  • Provides access to digital PDF content, such as: independent investigation inquiry suggestions; background information including content tables and concept; animations; PowerPoint™ concept presentation (and video version); comprehensive glossary
  • Special typewriter functionality in student activity PDFs allows students to take notes, record experimental data, answer analysis questions and e-mail their teacher
  • Correlated to the Next Generation Science Standards

Specifications

Grade 6 to 12
For Use With (Application) To introduces students the chemistry behind plant pigments (the blueberry biopigment cyanidan) and their use as ph indicators and fabric dyes
Includes Materials for three detailed activities (with model and inquiry investigational paths - a total of four investigational activities), which can accommodate up to 40 Students, DVD with PDF teacher and student guides, Structured, guided and open investigations, at introductory, intermediate and advanced levels
Class Size 40 Students
Product Type Ph, Indicators and Dyes Stem Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.