Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pH Neutralized IPA, Reagents

Catalog No. 01334956
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-956 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-956 Supplier Reagents Holdings Llc Supplier No. CI00140020F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

pH Neutralized IPA, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-63-0 , 77-09-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless to Pink
Quantity 20 L
Packaging Cubitainer
Physical Form Liquid
Specific Gravity 0.7808
Boiling Point 82.5°C
Grade Reagent
Chemical Name or Material pH Neutralized IPA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.