Learn More
Description
- Convenient dispenser allows you to select the right amount of pH paper for each application
- Easy-to-usedash (em)Dip paper in solution to be tested and match to color chart immediately
- Highly accurate and well suited for most applications
- Each SKU is packed in box of five clear plastic dispensers
- Cat. No. 13-640-507 is a pH paper testing set that includes shrink wrapped pack of seven dispensers (one of each SKU)
- Store at room temperature. Keep out of direct sunlight.
Order Info
15 ft. Roll provides about 100 tests.
Specifications
Specifications
| pH Range | 1 to 12 |
| For Use With (Application) | Convenient dispenser allows to select the right amount of pH paper for each application, highly accurate and well suited for most applications |
| Graduations | 1 |
| Quantity | 100 Tests |
| Storage Requirements | Room Temperature |
| Type | pH Test Paper |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
