Learn More
SPEX CERTIPREP LLC 3P pH 7 QC Check CRM, 500 mL
Supplier:  SPEX CERTIPREP LLC 3P QCI1127
Encompass_Preferred
(NSI by Zeptometrix) A 500 mL Ready-To-Use Bottle directly traceable to NIST SRM 187e.
Catalog No. NC0844722
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.