Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ pH Sensor
SDPSCI_ED

Catalog No. s16238nd Shop All Vernier Software Products
Change view
Click to view available options
Temperature Range (Metric):
5°C to 80°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
S16238ND 5°C to 80°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16238ND Supplier Vernier™ Supplier No. PHBTA
Only null left
Add to Cart
Add to Cart

Includes

Vernier pH Sensor, pH storage solution, pH storage solution bottle

Requires

Requires an interface and software, sold separately

Our general-purpose pH Sensor can be used across disciplines, including chemistry, biology, middle school science, and environmental science.

Type: Sealed, gel-filled, epoxy body, Ag/AgCl Shaft Diameter: 12 mm OD Response time: 90% of final reading in 1 second Temperature range: 5 to 80°C Range: pH 0–14 Accuracy: +/- 0.2 pH units Isopotential pH: pH 7 (point at which temperature has no effect)

Warranty and Services

5-year limited warranty

Specifications

Type Sealed, Gel-filled, Epoxy Body, Ag/AgCl Reference
Includes pH Sensor, pH storage solution, pH storage solution bottle
Grade Middle School, High School, College
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Accuracy ±0.2 pH units
Range pH 0 to 14
Response Time 90% of final reading in 1 second
Temperature Range (Metric) 5°C to 80°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.