Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ pH Test Strips
SCI_ED

Catalog No. S27896 Shop All Eisco Products
Change view
Click to view available options
pH Range:
2.8 to 4.4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
S27896 2.8 to 4.4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27896 Supplier Eisco™ Supplier No. WINETEST
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

pH test strips for detecting pH ranging from 2.8 to 4.4.

  • Measures pH ranging from 2.8 to 4.4
  • Water resistant color chart on vial for comparison
  • Strip Dimensions: 2.5 x 0.2 in.
  • 100 Strips/Vial

Specifications

pH Range 2.8 to 4.4
For Use With (Application) pH Testing and Monitoring
Includes Water Resistant Color Chart
Quantity 100 Pk.
Type Test Strip
Length (English) 2.5 in.
Width (English) 0.20 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.