Learn More
pH in Water- WP, Ready to use Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Packaging | Ampule |
| CAS | 1330-43-4 |
| Synonym | pH aq. soln.- WP |
| Type | Proficiency Testing Material |
| Quantity | 100 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.