Learn More
ILEX LIFE SCIENCES LLC PHA-L LYOPHILIZED 6 X 5ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0501010
Encompass_Preferred
PAN-Biotech Phytohemagglutinin-L (PHA-L), 6 x 5 ml (1.2 mg protein/flask), lyophilized. DESCRIPTION: PHA-L purified from Phaseolus vulgaris (red kidney bean). The lectin recognizes and binds specifically to terminal galactose, N-acetylglucosamine and mannose residues of complex glycans on mammalian glycoproteins. PHA-L is also known as leucoagglutinin and has high mitogenic and leucoagglutinating activity, but low erythroagglutinating activity. PHA-L does not agglutinate human erythrocytes at concentrations of 250 µg/mL or less, and is non-specific for blood groups. PHA-L lectin is supplied as a white lyophilized powder from a buffer containing from 10mM NH4HCO3. APPLICATION: Plant lectin suitable for stimulating lymphocyte proliferation and differentiation. Reconstitute one vial aseptically with 5 ml sterile pure water. Apply 1 ml of reconstituted solution to 100 ml cell culture medium. CAS REGISTRY NO.: 9008-97-3. STERILE: Yes. MANUFACTURER: PAN-Biotech GmbH (Germany).
Catalog No. 50-273-3748
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.