Learn More
Thermo Scientific™ Phadia™ Cardiolipin Antibodies for Antiphospholipid Syndrome
Catalog No. 14552901P
Change view
Click to view available options
Type:
EliA Cardiolipin IgA
EliA Cardiolipin IgG
EliA Cardiolipin IgM
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-552-901P EliA Cardiolipin IgG
14-552-801P EliA Cardiolipin IgA
14-553-001P EliA Cardiolipin IgM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 14-552-901P Supplier Thermo Scientific™ Supplier No. 14552901
Add to Cart
Add to Cart
This item is not returnable. View return policy
If present in the patient's specimen, antibodies to cardiolipin bind to their specific antigen. After washing away non-bound antibodies, enzyme-labeled antibodies against human IgG (IgM,IgA) antibodies (EliA IgG, IgM, IgA Conjugate™) are added to form an antibody-conjugate complex. After incubation, non-bound conjugate is washed away and the bound complex is incubated with a Development Solution. After stopping the reaction, the fluorescence in the reaction mixture is measured.To evaluate test results, the response for patient samples is compared directly to the response for calibrators.
  • The EliA Cardiolipin IgG (IgM, IgA) Wells are coated with bovine cardiolipin antigen
  • The higher the response value, the more specific IgG (IgM, IgA) is present in the specimen
  • All conjugates are isotype-specific -- mixed conjugate is not available
TRUSTED_SUSTAINABILITY
Specifications
Type EliA Cardiolipin IgG