Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Phagocytosis Assay Kit, Fluorescent, Red E. Coli, 100 Tests

Catalog No. 502586483
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-6483 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-6483 Supplier Abcam Supplier No. AB235901100TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Complete macrophage phagocytosis assay using fluorescent red-labeled E. coli suitable for microscopy, plate reader, or flow cytometry analysis.

  • Complete phagocytosis assay using fluorescent red-labeled E. coli
  • Suitable for analysis by microscope, plate reader, or flow cytometry
  • Compatible with both suspension and adherent cells

The product is subject to the following: Abcam Restricted Use Statement

Specifications

For Use With (Equipment) Microscope, Plate Reader, Flow Cytometer
Format Kit
No. of Reactions 100 Tests
Reactivity E. coli
Target Molecule E. coli
Detection Method Fluorescence
Sample Type Suspension cells, Adherent cells
Quantity 100 Tests
For Use With (Application) Phagocytosis assay, Microscopy, Plate reader, Flow cytometry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.